missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine Thrombin Protein
A cDNA sequence encoding the Thrombin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10878-2000U
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Thrombin Protein | |
Research Use Only | |
One unit is defined as the amount of enzyme needed to cleave 1 mg of fusion protein in 16 hours to 95% completion at 20C in a buffer containing 25mM Tris-HCl, pH 8.4, 150mM NaCl, and 2.5mM CaCl2. | |
Porcine | |
Untagged |
2000 IU | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Porcine | |
Lyophilized Powder from 20mM PBS, pH 7.4 containing 0.9% NaCl. |