missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Prolactin / PRL Protein
A DNA sequence encoding the Sus scrofa (Pig) Prolactin / PRL, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6531-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_999091.1 | |
Prolactin / PRL Protein | |
Research Use Only | |
PRL | |
Recombinant Protein | |
E. coli | |
LPICPSGAVN CQVSLRDLFD RAVILSHYIH NLSSEMFNEF DKRYAQGRGF ITKAINSCHT SSLSTPEDKE QAQQIHHEVL LNLILRVLRS WNDPLYHLVT EVRGMQEAPD AILSRAIEIE EQNKRLLEGM EKIVGQVHPG IKENEVYSVW SGLPSLQMAD EDTRLFAFYN LLHCLRRDSH KIDNYLKLLK CRIIYDSNC | |
Greater than 90% as determined by SDS-PAGE. |
396965 | |
200 μg | |
Not determined. | |
Full Length | |
Pig | |
His-SUMO | |
Tris-based buffer, 50% glycerol |