missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. aeruginosa Regulatory protein RhlR Protein
A DNA sequence encoding the Pseudomonas aeruginosa Regulatory protein RhlR, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7228-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_252167.1 | |
P. aeruginosa Regulatory protein RhlR Protein | |
Research Use Only | |
rhlR | |
Recombinant Protein | |
E. coli | |
MRNDGGFLLW WDGLRSEMQP IHDSQGVFAV LEKEVRRLGF DYYAYGVRHT IPFTRPKTEV HGTYPKAWLE RYQMQNYGAV DPAILNGLRS SEMVVWSDSL FDQSRMLWNE ARDWGLCVGA TLPIRAPNNL LSVLSVARDQ QNISSFEREE IRLRLRCMIE LLTQKLTDLE HPMLMSNPVC LSHREREILQ WTADGKSSGE IAIILSISES TVNFHHKNIQ KKFDAPNKTL AAAYAAALGL I | |
Greater than 84.5% as determined by SDS-PAGE. |
878968 | |
50 μg | |
Not determined. | |
Full Length | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |