missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Urease Protein
A cDNA sequence encoding the Urease was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10907-5mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Urease Protein | |
Research Use Only | |
The activity was found to be 141U/mg powder. | |
Other | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |
5 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
Each mg of protein contains 420microg Potassium Phosphate and 30microg EDTA Na2. |