missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other TEV Protein
A cDNA sequence encoding the TEV was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13725-1,000IU
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TEV Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
GHIVWPDYAN ILKEVFGGAR MACVTSAHMA GANGSILKKA EETSRATMHK PVIFGEDYVT EADLPYTPLH LEVNAEMERM YYLGRRALTH GKRRKVSVNN KRNRRRKVAK TYVGRDSIVE KIVVPHTERK VDTTTAVKDT CNEVSTQLVH NSMPKRKKQK NFLPATSLSN VYAQTWSIVR KRHMQVEIIS KKSVRAKVKR FEGSVQLFAS VRHMYGERKR VDLRIDNWQQ KTLLDLAKRF KNERVDQSKL TFGSSGLVLR QGSYAPAHWY RHGMFIVRGR SDGMLVDARA KVTFAVCYSM THY | |
Greater than 90.0% as determined by analysis by SDS-PAGE. |
1000 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
The rTEV contains 25mM Tris, 75mM NaCl, 5mM EDTA, 10mM GSH, 50% Glycerol. |