missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other MMLV RT Protein
A cDNA sequence encoding the MMLV RT was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12705-2500U
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MMLV RT Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, 0.1M NaCl, 0.1% Triton X-100, 2mM DTT, 0.1mM EDTA and 50% glycerol. |
2500 U | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |