missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Multi Species Vasopressin Protein
A cDNA sequence encoding the Vasopressin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13923-100mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
551 | |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
C[Cys-Tyr-Phe-Gln-Asn-Cys]-Pro-Arg-Gly-NH2 | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Multi Species Vasopressin Protein | |
Research Use Only | |
AVP | |
Multi Species | |
Untagged | |
The protein (1 mg/ml) was lyophilized with no additives. |