missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse VEGF Protein

A cDNA sequence encoding the VEGF was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10910-EC-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15985538

  • 5325.00€ / 1 miligramo
Fecha estimada de envío: 11-07-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Especificaciones

Especificaciones

VEGF Protein
Research Use Only
The ED50 range, determined by the dose-dependent proliferation of human umbilical vein endothelial cells (HUVEC) (measured by 3H-thymidine uptake) is 1-2 ng/ml, corresponding to a specific activity of 1x106 Units/mg.
Mouse
His
The protein was lyophilized from a concentrated (1 mg/ml) solution with no additives.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MNFLLSWVHW TLALLLYLHH AKWSQAAPTT EGEQKSHEVI KFMDVYQRSY CRPIETLVDI FQEYPDEIEY IFKPSCVPLM RCAGCCNDEA LECVPTSESN ITMQIMRIKP HQSQHIGEMS FLQHSRCECR PKKDRTKPEK KSVRGKGKGQ KRKRKKSRFK SWSVHCEPCS ERRKHLFVQD PQTCKCSCKN TDSRCKARQL ELNERTCRCD KPRR
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales