missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VAMP2 Protein
A cDNA sequence encoding the VAMP2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13916-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
VAMP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSATAA TVPPAAPAGE GGPPAPPPNL TSNRRLQQTQ AQVDEVVDIM RVNVDKVLER DQKLSELDDR ADALQAGASQ FETSAAKLKR KYWWKNLK | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
VAMP2 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 1mM EDTA, 0.1mM PMSF and 10% glycerol. |