missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Tryptase beta-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Tryptase beta-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6833-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Tryptase beta-2 Protein | |
Research Use Only | |
TPSB2 | |
Recombinant Protein | |
E. coli | |
IVGGHEASES KWPWQVSLRF KLNYWIHFCG GSLIHPQWVL TAAHCVGPHI KSPQLFRVQL REQYLYYGDQ LLSLNRIVVH PHYYTAEGGA DVALLELEVP VNVSTHIHPI SLPPASETFP PGTSCWVTGW GDIDNDEPLP PPYPLKQVKV PIVENSLCDR KYHTGLYTGD DFPIVHDGML CAGNTRRDSC QGDSGGPLVC KVKGTWLQAG VVSWGEGCAQ PNKPGIYTRV TYYLDWIHRY VPEHS | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |