missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Thrombospondin-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Thrombospondin-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6786-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_035711.2 | |
Thrombospondin-2 Protein | |
Research Use Only | |
Thbs2 | |
Recombinant Protein | |
E. coli | |
GDHVKDTSFD LFSISNINRK TIGAKQFRGP DPGVPAYRFV RFDYIPPVNT DDLNRIVKLA RRKEGFFLTA QLKQDRKSRG TLLVLEGPGT SQRQFEIVSN GPGDTLDLNY WVEGNQHTNF LEDVGLADSQ WKNVTVQVAS DTYSLYVGCD LIDSVTLEEP FYEQLEVDRS RMYVAKGASR ESHFRGLLQN VHLVFADSVE DILSKKGCQH SQGA | |
Greater than 90% as determined by SDS-PAGE. |
21826 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |