missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Thrombopoietin / THPO Protein
A DNA sequence encoding the Mouse Thrombopoietin / THPO, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10468-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001166976.1 | |
Thrombopoietin / THPO Protein | |
Research Use Only | |
THPO | |
The ED50 as determined by a cell proliferation assay using human MO7e cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, with 5% Trehalose, lyophilized |
21832 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SPVAPACDPR LLNKLLRDSH LLHSRLSQCP DVDPLSIPVL LPAVDFSLGE WKTQTEQSKA QDILGAVSLL LEGVMAARGQ LEPSCLSSLL GQLSGQVRLL LGALQGLLGT QLPLQGRTTA HKDPNALFLS LQQLLRGKVR FLLLVEGPTL CVRRTLPTTA VPSSTSQLLT LNKF | |
Greater than 95% as determined by SDS-PAGE and HPLC. |