Learn More
enQuireBio™ Recombinant Mouse sRANKL Protein
A cDNA sequence encoding the sRANKL was constructed and used to recombinantly synthesize the protein.
Marca:  enQuireBio™ QP10869-1mg
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15925418
  • 7090.00€ / 1 miligramo
Fecha estimada de envío: 11-07-2024
para ver el stock
Especificaciones
sRANKL Protein
Research Use Only
The ED50, as measured by its ability to induce osteoclast differentiation of RAW 264.7 mouse monocyte/macrophage cells, is less than 2ng/ml.
Mouse
Untagged
sRANKL protein was lyophilized from a 0.2microm filtered solution in NaH2PO4, NaCl and EDTA.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
Lys158-Asp316, with an N-terminal Met, Accession # Q3TWY5
Greater than 95.0% as determined by SDS-PAGE.