missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Small proline-rich protein 2A Protein
A DNA sequence encoding the Mus musculus (Mouse) Small proline-rich protein 2A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6723-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001158259.1 | |
Small proline-rich protein 2A Protein | |
Research Use Only | |
Sprr2a | |
Recombinant Protein | |
E. coli | |
MSYYQQQCNQ PCRPPPVCPP PKCPEPCPPQ VWPGPCRPVM CFEPCLPSVW PGPCRPVVCY EQCPPQPWQS TCPPVQFPPC QQK | |
Greater than 90% as determined by SDS-PAGE. |
100042514 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |