missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Selenoprotein 1
A DNA sequence encoding the Mus musculus (Mouse) Selenoprotein T, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7250-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001035486.2 | |
Selenoprotein 1 | |
Research Use Only | |
Selt | |
Recombinant Protein | |
E. coli | |
SANLGGVPSK RLKMQYATGP LLKFQICVSU GYRRVFEEYM RVISQRYPDI RIEGENYLPQ PIYRHIASFL SVFKLVLIGL IIVGKDPFAF FGMQAPSIWQ WGQENKVYAC MMVFFLSNMI ENQCMSTGAF EITLNDVPVW SKLESGHLPS MQQLVQILDN EMKLNVHMDS IPHHRS | |
Greater than 90% as determined by SDS-PAGE. |
69227 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |