missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Platelet-derived growth factor D Protein
A DNA sequence encoding the Mus musculus (Mouse) Platelet-derived growth factor D, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7981-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_082200.1 | |
Platelet-derived growth factor D Protein | |
Research Use Only | |
Pdgfd | |
Recombinant Protein | |
E. coli | |
TPQRASIKAL RNANLRRDES NHLTDLYQRE ENIQVTSNGH VQSPRFPNSY PRNLLLTWWL RSQEKTRIQL SFDHQFGLEE AENDICRYDF VEVEEVSESS TVVRGRWCGH KEIPPRITSR TNQIKITFKS DDYFVAKPGF KIYYSFVEDF QPEAASETNW ESVTSSFSGV SYHSPSITDP TLTADALDKT VAEFDTVEDL LKHFNPVSWQ DDLENLYLDT PHYRGRSYHD RKSKVDLDRL NDDVKRYSCT PRNHSVNLRE ELKLTNAVFF PRCLLVQRCG GNCGCGTVNW KSCTCSSGKT VKKYHEVLKF EPGHFKRRGK AKNMALVDIQ LDHHERCDCI CSSRPPR | |
Greater than 80% as determined by SDS-PAGE. |
71785 | |
1 mg | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |