missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MIF / GLIF Protein
A DNA sequence encoding the Mouse MIF / GLIF, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP1493-25 ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_034928.1 | |
MIF / GLIF Protein | |
Research Use Only | |
MIF | |
Mouse | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
17319 | |
25 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Recombinant Protein | |
E. coli | |
MPMFIVNTNV PRASVPEGFL SELTQQLAQA TGKPAQYIAV HVVPDQLMTF SGTNDPCALC SLHSIGKIGG AQNRNYSKLL CGLLSDRLHI SPDRVYINYY DMNAANVGWN GSTFA | |
Greater than 96% as determined by SDS-PAGE and HPLC. |