missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse L-FABP / FABP1 Protein
A DNA sequence encoding the Mouse L-FABP / FABP1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10197-ec-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_059095.1 | |
L-FABP / FABP1 Protein | |
Research Use Only | |
Fabp1 | |
The binding affinity of rMuFABP1 for the synthetic ligand cis-parinaric acid has been measured by fluorescence titration. Half maximal fluorescence of 2.5 uM rMuFABP1 is achieved with approximately 5 uM cis-paranaric acid. | |
Mouse | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
14080 | |
25 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MNFSGKYQLQ SQENFEPFMK AIGLPEDLIQ KGKDIKGVSE IVHEGKKIKL TITYGPKVVR NEFTLGEECE LETMTGEKVK AVVKLEGDNK MVTTFKGIKS VTELNGDTIT NTMTLGDIVY KRVSKRI | |
Greater than 95% as determined by SDS-PAGE and HPLC. |