missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Interferon regulatory factor 8 Protein
A DNA sequence encoding the Mus musculus (Mouse) Interferon regulatory factor 8, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6235-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001288740.1 | |
Interferon regulatory factor 8 Protein | |
Research Use Only | |
Irf8 | |
Recombinant Protein | |
E. coli | |
MCDRNGGRRL RQWLIEQIDS SMYPGLIWEN DEKTMFRIPW KHAGKQDYNQ EVDASIFKAW AVFKGKFKEG DKAEPATWKT RLRCALNKSP DFEEVTDRSQ LDISEPYKVY RIVPEEEQKC KLGVAPAGCM SEVPEMECGR SEIEELIKEP SVDEYMGMTK RSPSPPEACR SQILPDWWVQ QPSAGLPLVT GYAAYDTHHS AFSQMVISFY YGGKLVGQAT TTCLEGCRLS LSQPGLPKLY GPDGLEPVCF PTADTIPSER QRQVTRKLFG HLERGVLLHS NRKGVFVKRL CQGRVFCSGN AVVCKGRPNK LERDEVVQVF DTNQFIRELQ QFYATQSRLP DSRVVLCFGE EFPDTVPLRS KLILVQVEQL YARQLVEEAG KSCGAGSLMP ALEEPQPDQA FRMFPDICTS HQRPFFRENQ QITV | |
Greater than 90% as determined by SDS-PAGE. |
15900 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |