missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL36RN Protein
A cDNA sequence encoding the IL36RN was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10730-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
IL36RN Protein | |
Research Use Only | |
Fully biologically active when compared to standard. Measured by its ability to inhibit IL-36a, IL-36b or IL-36g induced IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells. The ED50 for this effect is typically 0.8-4ug/ml (corresponding to a specific activity of 250-125units/mg) in the presence of 15ng/ml of recombinant mouse IL-36b. | |
Mouse | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
VLSGALCFRM KDSALKVLYL HNNQLLAGGL HAEKVIKGEE ISVVPNRALD ASLSPVILGV QGGSQCLSCG TEKGPILKLE PVNIMELYLG AKESKSFTFY RRDMGLTSSF ESAAYPGWFL CTSPEADQPV RLTQIPEDPA WDAPITDFYF QQCD | |
Greater than 95.0% as determined by SDS-PAGE. |