Learn More
enQuireBio™ Recombinant Mouse IL36RN Protein
A cDNA sequence encoding the IL36RN was constructed and used to recombinantly synthesize the protein.
137.00€ - 7090.00€
Especificaciones
Nombre IL36RN Protein
Estado normativo Research Use Only
Concentración de endotoxinas < 1.0 EU per ug protein as determined by the LAL method.
Actividad biológica Fully biologically active when compared to standard. Measured by its ability to inhibit IL-36a, IL-36b or IL-36g induced IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells. The ED50 for this effect is typically 0.8-4ug/ml (corresponding to a specific activity of 250-125units/mg) in the presence of 15ng/ml of recombinant mouse IL-36b.
Tipo de producto Recombinant Protein
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15914998
Ver documentos Detalles de la promoción
enQuireBio™
QP10730-2UG
2 μg
137.00€
2 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 29-05-2024
para ver el stock
15994988
Ver documentos Detalles de la promoción
enQuireBio™
QP10730-10UG
10 μg
212.00€
10 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 29-05-2024
para ver el stock
15904998
Ver documentos Detalles de la promoción
enQuireBio™
QP10730-1MG
1 mg
7090.00€
1 miligramo
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 29-05-2024
para ver el stock
Especificaciones
IL36RN Protein
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
VLSGALCFRM KDSALKVLYL HNNQLLAGGL HAEKVIKGEE ISVVPNRALD ASLSPVILGV QGGSQCLSCG TEKGPILKLE PVNIMELYLG AKESKSFTFY RRDMGLTSSF ESAAYPGWFL CTSPEADQPV RLTQIPEDPA WDAPITDFYF QQCD
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Fully biologically active when compared to standard. Measured by its ability to inhibit IL-36a, IL-36b or IL-36g induced IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells. The ED50 for this effect is typically 0.8-4ug/ml (corresponding to a specific activity of 250-125units/mg) in the presence of 15ng/ml of recombinant mouse IL-36b.
Mouse
Untagged
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.