missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL21 / Interleukin 21 Protein
A DNA sequence encoding the Mouse IL21 / Interleukin 21, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10462-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_068554.1 | |
IL21 / Interleukin 21 Protein | |
Research Use Only | |
IL21 | |
The ED50 as determined by a cell proliferation assay using human N1186 T cells is less than 25 ng/ml, corresponding to a specific activity of > 4.0 M 104 IU/mg. | |
Mouse | |
Untagged | |
0.2 m filtered 2 M PBS, pH 7.4, lyophilized |
60505 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
HKSSPQGPDR LLIRLRHLID IVEQLKIYEN DLDPELLSAP QDVKGHCEHA AFACFQKAKL KPSNPGNNKT FIIDLVAQLR RRLPARRGGK KQKHIAKCPS CDSYEKRTPK EFLERLKWLL QKMIHQHLS | |
Greater than 98% as determined by SDS-PAGE and HPLC. |