missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL1F5 / IL36RN Protein
A DNA sequence encoding the Mouse IL1F5 / IL36RN, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10438-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001139559.1 | |
IL1F5 / IL36RN Protein | |
Research Use Only | |
IL36RN | |
Measured by its ability to inhibit IL-36gamma induced IL-6 production by human PBMCs. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
54450 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
VLSGALCFRM KDSALKVLYL HNNQLLAGGL HAEKVIKGEE ISVVPNRALD ASLSPVILGV QGGSQCLSCG TEKGPILKLE PVNIMELYLG AKESKSFTFY RRDMGLTSSF ESAAYPGWFL CTSPEADQPV RLTQIPEDPA WDAPITDFYF QQCD | |
Greater than 95% as determined by SDS-PAGE and HPLC. |