missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL 28B Protein
A cDNA sequence encoding the IL 28B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10710-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
338374 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by an anti-viral assay using human HepG2 cells infected with encephalomyocarditis is less than 30 ng/ml, corresponding to a specific activity of > 3.3 M 104 IU/mg. | |
Mouse | |
Untagged | |
IL28B protein was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |
IL 28B Protein | |
Research Use Only | |
Ifnl3 | |
Recombinant Protein | |
E. coli | |
DPVPRATRLP VEAKDCHIAQ FKSLSPKELQ AFKKAKGAIE KRLLEKDMRC SSHLISRAWD LKQLQVQERP KALQAEVALT LKVWENINDS ALTTILGQPL HTLSHIHSQL QTCTQLQATA EPKPPSRRLS RWLHRLQEAQ SKETPGCLED SVTSNLFQLL LRDLKCVASG DQCV | |
Greater than 97.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |