missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-1RA / IL1RN Protein
A DNA sequence encoding the Mouse IL-1RA / IL1RN, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10450-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001034790.1 | |
IL-1RA / IL1RN Protein | |
Research Use Only | |
Il1rn | |
The ED50 as determined by inhibiting IL-1alpha- dependent proliferation of murine D10S cells is less than 50 ng/ml, corresponding to a specific activity of > 2.0 M 104 IU/mg in the presence of 50 pg/ml rHuIL-1alpha. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
16181 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
RPSGKRPCKM QAFRIWDTNQ KTFYLRNNQL IAGYLQGPNI KLEEKIDMVP IDLHSVFLGI HGGKLCLSCA KSGDDIKLQL EEVNITDLSK NKEEDKRFTF IRSEKGPTTS FESAACPGWF LCTTLEADRP VSLTNTPEEP LIVTKFYFQE DQ | |
Greater than 97% as determined by SDS-PAGE and HPLC. |