missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GDNF Protein
Recombinant Protein
Marca: enQuireBio™ QP5446-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001288261.1 | |
GDNF Protein | |
Research Use Only | |
GDNF | |
Recombinant Protein | |
E. coli | |
MSPDKQAALP RRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI | |
Greater than 95% as determined by SDS-PAGE. |
14573 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Bioactivity of recombinant GDNF is confirmed by C6 cell proliferation assay (see figure). | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |