Learn More
enQuireBio™ Recombinant Mouse GDNF Protein
Recombinant Protein
112.00€ - 7420.00€
Especificaciones
Número de acceso NP_001288261.1
ID de gen (Entrez) 14573
Nombre GDNF Protein
Estado normativo Research Use Only
Concentración de endotoxinas Less than or equal to 1.00 EUs/ug as determined by the LAL method.
View More Specs
Products 5
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
15996139
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP5446-2UG
2 μg
112.00€
2 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 21-06-2024
para ver el stock
15976139
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP5446-10UG
10 μg
263.00€
10 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 21-06-2024
para ver el stock
15966139
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP5446-100UG
100 μg
985.00€
100 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 21-06-2024
para ver el stock
15906149
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP5446-500UG
500 μg
4515.00€
500 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 21-06-2024
para ver el stock
15986139
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP5446-1MG
1 mg
7420.00€
1 miligramo
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 21-06-2024
para ver el stock
Description
Description
Specifications
Specifications
NP_001288261.1
GDNF Protein
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Bioactivity of recombinant GDNF is confirmed by C6 cell proliferation assay (see figure).
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
14573
Research Use Only
GDNF
Recombinant Protein
E. coli
MSPDKQAALP RRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI
Greater than 95% as determined by SDS-PAGE.
Videos
SDS
Documents
Documents
Certificates