missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GDF7 Protein
A cDNA sequence encoding the GDF7 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10629-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
238057 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by inducing alkaline phosphatase production of murine ATDC5 cells is less than 0.5microg/ml, corresponding to a specific activity of > 2000 IU/mg. | |
Mouse | |
Untagged | |
GDF7 protein was lyophilized from a 0.2microm filtered concentrated solution in 30% Acetonitrile and 0.1% TFA. |
GDF7 Protein | |
Research Use Only | |
Gdf7 | |
Recombinant Protein | |
E. coli | |
TALAGTRGAQ GSGGGGGGGG GGGGGGGGGG GGAGRGHGRR GRSRCSRKSL HVDFKELGWD DWIIAPLDYE AYHCEGVCDF PLRSHLEPTN HAIIQTLLNS MAPDAAPASC CVPARLSPIS ILYIDAANNV VYKQYEDMVV EACGCR | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |