Learn More
enQuireBio™ Recombinant Mouse Follistatin / FST Protein
Recombinant Protein
Marca:  enQuireBio™ QP5428-1mg
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15996099
  • 7420.00€ / 1 miligramo
Fecha estimada de envío: 27-06-2024
para ver el stock
Especificaciones
NP_001288302.1
Follistatin / FST Protein
Research Use Only
FST
Recombinant Protein
E. coli
MGNCWLRQAK NGRCQVLYKT ELSKEECCST GRLSTSWTEE DVNDNTLFKW MIFNGGAPNC IPCKETCENV DCGPGKKCRM NKKNKPRCVC APDCSNITWK GPVCGLDGKT YRNECALLKA RCKEQPELEV QYQGKCKKTC RDVFCPGSST CVVDQTNNAY CVTCNRICPE PSSSEQYLCG NDGVTYSSAC HLRKATCLLG RSIGLAYEGK CIKAKSCEDI QCGGGKKCLW DSKVGRGRCS LCDELCPDSK SDEPVCASDN ATYASECAMK EAACSSGVLL EVKHSGSCN
Greater than 90 as determined by SDS-PAGE.
14313
1 mg
Less than or equal to 5.00 EUs/ug as determined by the LAL method.
Mouse Follistatin / FST induces MCP-11 cell proliferation (typical ED50 ? 1 ug/ml).
Mouse
Untagged
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5