missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse FGF 18 Protein
A cDNA sequence encoding the FGF 18 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10597-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FGF 18 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
EENVDFRIHV ENQTRARDDV SRKQLRLYQL YSRTSGKHIQ VLGRRISARG EDGDKYAQLL VETDTFGSQV RIKGKETEFY LCMNRKGKLV GKPDGTSKEC VFIEKVLENN YTALMSAKYS GWYVGFTKKG RPRKGPKTRE NQQDVHFMKR YPKGQAELQK PFKYTTVTKR SRRIRPTHPG | |
Greater than 95.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Mouse | |
Untagged | |
FGF-18 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |