missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL5 / ENA-78 Protein
A DNA sequence encoding the Mouse CXCL5 / ENA-78, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10253-ec-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_033167.2 | |
CXCL5 / ENA-78 Protein | |
Research Use Only | |
CXCL5 | |
The biological activity determined by a chemoattract bioassay using human neutrophils is in a concentration range of 10-100 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
20311 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
TELRCVCLTV TPKINPKLIA NLEVIPAGPQ CPTVEVIAKL KNQKEVCLDP EAPVIKKIIQ KILGSDKKKA | |
Greater than 96% as determined by SDS-PAGE and HPLC. |