missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL10 / Crg-2 Protein
Recombinant Protein
Marca: enQuireBio™ QP5448-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_067249.1 | |
CXCL10 / Crg-2 Protein | |
Research Use Only | |
CXCL10 | |
Recombinant Protein | |
E. coli | |
IPLARTVRCN CIHIDDGPVR MRAIGKLEII PASLSCPRVE IIATMKKNDE QRCLNPESKT IKNLMKAFSQ KRSKRAP | |
Greater than 95% as determined by SDS-PAGE. |
15945 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse CXCL10 / Crg-2 induces Primary human T cell chemotaxis (significanty different from control at Mouse CXCL10 / Crg-2 concentrations greater than 5 ng/ml). | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |