missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL10 / Crg-2 Protein
A DNA sequence encoding the Mouse CXCL10 / Crg-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10256-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Specifications
NP_067249.1 | |
CXCL10 / Crg-2 Protein | |
Research Use Only | |
CXCL10 | |
The biologically active determined by a chemotaxis bioassay using human peripheral blood lymphocytes is in a concentration range of 0.1- 10.0 ng/ml in the presence of IL-2. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
15945 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
IPLARTVRCN CIHIDDGPVR MRAIGKLEII PASLSCPRVE IIATMKKNDE QRCLNPESKT IKNLMKAFSQ KRSKRAP | |
Greater than 97% as determined by SDS-PAGE and HPLC. |