missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD36 Protein
A cDNA sequence encoding the CD36 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11329-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
12491 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPGDMLIEK TIKREVVLEE GTTAFKNWVK TGTTVYRQFW IFDVQNPDDV AKNSSKIKVK QRGPYTYRVR YLAKENITQD PEDHTVSFVQ PNGAIFEPSL SVGTEDDNFT VLNLAVAAAP HIYQNSFVQV VLNSLIKKSK SSMFQTRSLK ELLWGYKDPF LSLVPYPIST TVGVFYPYND TVDGVYKVFN GKDNISKVAI IESYKGKRNL SYWPSYCDMI NGTDAASFPP FVEKSRTLRF FSSDICRSIY AVFGSEIDLK GIPVYRFVLP ANAFASPLQN PDNHCFCTEK VISNNCTSYG VLDIGKCKEG KPVYISLPHF LHASPDVSEP IEGLHPNEDE HRTYLDVEPI TGFTLQFAKR LQVNILVKPA RKIEALKNLK RPYIVPILWL NETGTIGDEK AEMFKTQVTG KIKHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
CD36 Protein | |
Research Use Only | |
Cd36 | |
Mouse | |
His | |
CD36 protein solution (1 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |