missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse C-C motif chemokine 25 Protein
A DNA sequence encoding the Mouse C-C motif chemokine 25, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10278-ec-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_033164.1 | |
C-C motif chemokine 25 Protein | |
Research Use Only | |
CCL25 | |
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 5.0-50 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
20300 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
QGAFEDCCLG YQHRIKWNVL RHARNYHQQE VSGSCNLRAV RFYFRQKVVC GNPEDMNVKR AIRILTARKR LVHWKSASDS QTERKKSNHM KSKVENPNST SVRSATLGHP RMVMMPRKTN N | |
Greater than 95% as determined by SDS-PAGE and HPLC. |