missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Beclin-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Beclin-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5715-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_062530.2 | |
Beclin-1 Protein | |
Research Use Only | |
Becn1 | |
Recombinant Protein | |
E. coli | |
MEGSKASSST MQVSFVCQRC SQPLKLDTSF KILDRVTIQE LTAPLLTTAQ AKPGETQEEE ANSGEEPFIE TRQDGVSRRF IPPARMMSTE SANSFTLIGE ASDGGTMENL SRRLKVTGDL FDIMSGQTDV DHPLCEECTD TLLDQLDTQL NVTENECQNY KRCLEILEQM NEDDSEQLQR ELKELALEEE RLIQELEDVE KNRKVVAENL EKVQAEAERL DQEEAQYQRE YSEFKRQQLE LDDELKSVEN QVRYAQIQLD KLKKTNVFNA TFHIWHSGQF GTINNFRLGR LPSVPVEWNE INAAWGQTVL LLHALANKMG LKFQRYRLVP YGNHSYLESL TDKSKELPLY CSGGLRFFWD NKFDHAMVAF LDCVQQFKEE VEKGETRFCL PYRMDVEKGK IEDTGGSGGS YSIKTQFNSE EQWTKALKFM LTNLKWGLAW VSSQFYNK | |
Greater than 90% as determined by SDS-PAGE. |
56208 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |