missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Bcl XL Protein
A cDNA sequence encoding the Bcl XL was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11140-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Bcl XL Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
SQSNRELVVD FLSYKLSQKG YSWSQFSDVE ENRTEAPEET EAERETPSAI NGNPSWHLAD SPAVNGATGH SSSLDAREVI PMAAVKQALR EAGDEFELRY RRAFSDLTSQ LHITPGTAYQ SFEQVVNELF RDGVNWGRIV AFFSFGGALC VESVDKEMQV LVSRIASWMA TYLNDHLEPW IQENGGWDTF VDLYGNNAAA ESRKGQERFN R | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
Untagged | |
Bcl-XL Mouse was lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4 and 5% Trehalose. |