missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse APRIL Protein
A cDNA sequence encoding the APRIL was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10523-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
69583 | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Determined by a cell proliferation assay using activated T cells. | |
Mouse | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4, with 0.02 % Tween-20. |
APRIL Protein | |
Research Use Only | |
Tnfsf13 | |
Recombinant Protein | |
E. coli | |
AVLTQKHKKK HSVLHLVPVN ITSKADSDVT EVMWQPVLRR GRGLEAQGDI VRVWDTGIYL LYSQVLFHDV TFTMGQVVSR EGQGRRETLF RCIRSMPSDP DRAYNSCYSA GVFHLHQGDI ITVKIPRANA KLSLSPHGTF LGFVKL | |
Greater than 97.0% as determined by Analysis by SDS-PAGE. |