missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse APOA1 Protein
A cDNA sequence encoding the APOA1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11044-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
11806 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSDEPQSQW DKVKDFANVY VDAVKDSGRD YVSQFESSSL GQQLNLNLLE NWDTLGSTVS QLQERLGPLT RDFWDNLEKE TDWVRQEMNK DLEEVKQKVQ PYLDEFQKKW KEDVELYRQK VAPLGAELQE SARQKLQELQ GRLSPVAEEF RDRMRTHVDS LRTQLAPHSE QMRESLAQRL AELKSNPTLN EYHTRAKTHL KTLGEKARPA LEDLRHSLMP MLETLKTQVQ SVIDKASETL TAQ | |
Greater than 90.0% as determined by SDS-PAGE. |
APOA1 Protein | |
Research Use Only | |
Apoa1 | |
Mouse | |
His | |
APOA1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |