missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL6 / Interleukin-6 Protein
A DNA sequence encoding the Rhesus Macaque IL6 / Interleukin-6 Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP1556-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
IL6 / Interleukin-6 Protein | |
Research Use Only | |
IL6 | |
The ED50 as determined by a cell proliferation assay usingIL-6-dependent murine 7TD1 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg. | |
Macaque | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MAPVLPGEDS KNVAAPHSQP LTSSERIDKH IRYILDGISA LRKETCNRSN MCESSKEALA ENNLNLPKMA EKDGCFQSGF NEDTCLVKII TGLLEFEVYL EYLQNRFESS EEQARAVQMS TKVLIQFLQK KAKNLDAITT PEPTTNASLL TKLQAQNQWL QDMTTHLILR SFKEFLQSNL RALRQM | |
Greater than 97% as determined by SDS-PAGE and HPLC. |