missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL-8 / CXCL8 Protein
A DNA sequence encoding the Rhesus Macaque IL-8 / CXCL8, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP1557-ec-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001028137.1 | |
IL-8 / CXCL8 Protein | |
Research Use Only | |
CXCL8 | |
The biological activity determined by a chemotaxis bioassay using human CXCR2 transfected murine BaF3 cells is in a concentration range of 0.5-5.0 ng/ml. | |
Macaque | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
613028 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
AVLPRSAKEL RCECIKTYSK PFHPKFIKEL RVIESGPHCA NTEIIVKLSD GRELCLDPKE PWVQRVVEKF VKRAENQNP | |
Greater than 95% as determined by SDS-PAGE and HPLC. |