missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL-1 beta / IL1B Protein
A DNA sequence encoding the Rhesus Macaque IL-1 beta / IL1B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10434-ec-10ug
Additional Details : Peso : 0.01000kg
Specifications
NP_001036221.1 | |
IL-1 beta / IL1B Protein | |
Research Use Only | |
IL1B | |
The ED50 as determined by a cell proliferation assay using murine D10.G4.1 cells is less than 10 pg/ml, corresponding to a specific activity of > 1.0 M 108 IU/mg. | |
Macaque | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
704701 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APVRSLHCTL RDAQLKSLVM SGPYELKALH LQGQDLEQQV VFSMSFVQGE ESNDKIPVAL GLKAKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTRGG QDITDFTMQF VSS | |
Greater than 98% as determined by SDS-PAGE and HPLC. |