missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque CXCL10 / Crg-2 Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) CXCL10 / Crg-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7849-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001028064.1 | |
CXCL10 / Crg-2 Protein | |
Research Use Only | |
CXCL10 | |
Recombinant Protein | |
E. coli | |
IPLSRTVRCT CISISNQPVN PRSLEKLEII PPSQFCPHVE IIATMKKKGE KRCLNPESKA IKNLLKAVSK ERSKRSP | |
Greater than 90% as determined by SDS-PAGE. |
574243 | |
100 μg | |
Not determined. | |
Full Length | |
Macaque | |
His | |
Tris-based buffer, 50% glycerol |