Learn More
enQuireBio™ Recombinant Japanese encephalitis Virus- NS1 Protein
Recombinant Protein
Marca: enQuireBio™ QP10487-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Japanese encephalitis Virus- NS1 Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
DPBS pH 7.4. |
100 μg | |
Untested | |
Japanese encephalitis virus | |
His | |
>95% as determined by SDS-PAGE. |