missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZSCAN21 Protein
A DNA sequence encoding the Homo sapiens (Human) ZSCAN21, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8774-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_666019.1 | |
ZSCAN21 Protein | |
Research Use Only | |
ZSCAN21 | |
Recombinant Protein | |
E. coli | |
MMTKVLGMAP VLGPRPPQEQ VGPLMVKVEE KEEKGKYLPS LEMFRQRFRQ FGYHDTPGPR EALSQLRVLC CEWLRPEIHT KEQILELLVL EQFLTILPQE LQAWVQEHCP ESAEEAVTLL EDLERELDEP GHQVSTPPNE QKPVWEKISS SGTAKESPSS MQPQPLETSH KYESWGPLYI QESGEEQEFA QDPRKVRDCR LSTQHEESAD EQKGSEAEGL KGDIISVIIA NKPEASLERQ CVNLENEKGT KPPLQEAGSK KGRESVPTKP TPGERRYICA ECGKAFSNSS NLTKHRRTHT GEKPYVCTKC GKAFSHSSNL TLHYRTHLVD RPYDCKCGKA FGQSSDLLKH QRMHTEEAPY QCKDCGKAFS GKGSLIRHYR IHTGEKPYQC NECGKSFSQH AGLSSHQRLH TGEKPYKCKE CGKAFNHSSN FNKHHRIHTG EKPYWCHHCG KTFCSKSNLS K | |
Greater than 90% as determined by SDS-PAGE. |
7589 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |