missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZG16 Protein
A cDNA sequence encoding the ZG16 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13995-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
653808 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSNAIQARS SSYSGEYGGG GGKRFSHSGN QLDGPITALR VRVNTYYIVG LQVRYGKVWS DYVGGRNGDL EEIFLHPGES VIQVSGKYKW YLKKLVFVTD KGRYLSFGKD SGTSFNAVPL HPNTVLRFIS GRSGSLIDAI GLHWDVYPTS CSRC | |
Greater than 95% as determined by SDS-PAGE. |
ZG16 Protein | |
Research Use Only | |
ZG16 | |
Human | |
His | |
The ZG16 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |