missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VEGF / VEGFA / VEGF165 Protein
A DNA sequence encoding the Human VEGF / VEGFA / VEGF165, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10395-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001020537.2 | |
VEGF / VEGFA / VEGF165 Protein | |
Research Use Only | |
VEGFA | |
The ED50 as determined by a cell proliferation assay using human umbilical vein endothelial cells(HUVEC) is between 1.0-8.0 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
7422 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MAPMAEGGGQ NHHEVVKFMD VYQRSYCHPI ETLVDIFQEY PDEIEYIFKP SCVPLMRCGG CCNDEGLECV PTEESNITMQ IMRIKPHQGQ HIGEMSFLQH NKCECRPKKD RARQENPCGP CSERRKHLFV QDPQTCKCSC KNTDSRCKAR QLELNERTCR CDKPRR | |
Greater than 95% as determined by SDS-PAGE and HPLC. |