Learn More
enQuireBio™ Recombinant Human UV excision repair protein RAD23 homolog A Protein
A DNA sequence encoding the Homo sapiens (Human) UV excision repair protein RAD23 homolog A, was expressed in the hosts and tags indicated.
Marca:  enQuireBio™ QP6578-ec-1mg
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 16044290
  • 2365.00€ / 1 miligramo
Fecha estimada de envío: 17-08-2022
para ver el stock
Especificaciones
NP_001257291.1
UV excision repair protein RAD23 homolog A Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
5886
Greater than 80% as determined by SDS-PAGE.
Research Use Only
RAD23A
Recombinant Protein
E. coli
MAVTITLKTL QQQTFKIRME PDETVKVLKE KIEAEKGRDA FPVAGQKLIY AGKILSDDVP IRDYRIDEKN FVVVMVTKTK AGQGTSAPPE ASPTAAPESS TSFPPAPTSG MSHPPPAARE DKSPSEESAP TTSPESVSGS VPSSGSSGRE EDAASTLVTG SEYETMLTEI MSMGYERERV VAALRASYNN PHRAVEYLLT GIPGSPEPEH GSVQESQVSE QPATEAAGEN PLEFLRDQPQ FQNMRQVIQQ NPALLPALLQ QLGQENPQLL QQISRHQEQF IQMLNEPPGE LADISDVEGE VGAIGEEAPQ MNYIQVTPQE KEAIERLKAL GFPESLVIQA YFACEKNENL AANFLLSQNF DDE