missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Uteroglobin / SCGB1A1 Protein
A DNA sequence encoding the Human Uteroglobin / SCGB1A1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10189-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_003348.1 | |
Uteroglobin / SCGB1A1 Protein | |
Research Use Only | |
SCGB1A1 | |
The ED50 as determined by the ability of the immobilized protein to support the adhesion of the A549 human lung carcinoma cells is less than 5.0 ug/ml, corresponding to a specific activity of > 200 IU/m. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
7356 | |
50 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
EICPSFQRVI ETLLMDTPSS YEAAMELFSP DQDMREAGAQ LKKLVDTLPQ KPRESIIKLM EKIAQSSLCN | |
Greater than 98% as determined by SDS-PAGE and HPLC. |