missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UPP1 Protein
A cDNA sequence encoding the UPP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13900-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
UPP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAATGANAEK AESHNDCPVR LLNPNIAKMK EDILYHFNLT TSRHNFPALF GDVKFVCVGG SPSRMKAFIR CVGAELGLDC PGRDYPNICA GTDRYAMYKV GPVLSVSHGM GIPSISIMLH ELIKLLYYAR CSNVTIIRIG TSGGIGLEPG TVVITEQAVD TCFKAEFEQI VLGKRVIRKT DLNKKLVQEL LLCSAELSEF TTVVGNTMCT LDFYEGQGRL DGALCSYTEK DKQAYLEAAY AAGVRNIEME SSVFAAMCSA CGLQAAVVCV TLLNRLEGDQ ISSPRNVLSE YQQRPQRLVS YFIKKKLSKA | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UPP1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.2M NaCl and 40% glycerol. |